missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ AAV-MAX Helper-Free AAV Production System Kit DFS Item

The Gibco AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production.

Supplier:  Gibco™ A51217

Catalog No. A51217


Explore available promotions

Description

Description

The Gibco AAV-MAX Helper-Free AAV Production System is based on a high-density suspension culture of clonal 293F-derived Viral Production Cells 2.0 in Viral Production Medium. Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel production enhancer. The components are designed to work synergistically, resulting in maximal titers and eliminating the need to optimize reagents and protocols.

Features of the AAV-MAX Helper-Free AAV Production System include:

  • Components work synergistically to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • Suspension-based system has scalable protocols from shake flask to bioreactor scale
  • Simplified workflow—streamlined protocol using helper virus-free triple transfection
  • Clonal, 293F-derived, high-producer cell line
  • Robust transfection protocols are adaptable to large-scale transfection

The components included in the AAV-MAX System Kit are:

  • 1 vial of Viral Production Cells 2.0
  • 1 L of Viral Production Medium
  • 1 AAV-MAX Transfection Kit for 1 L of production
  • 100 mL Viral-Plex Complexation Buffer
  • 100 mL AAV-MAX Lysis Buffer

All components are also available for purchase separately.

Specifications

Specifications

• 1 vial Viral Production Cells 2.0
• 1 L Viral Production Medium
• 1 AAV-MAX Transfection Kit for 1 L production
• 100 mL Viral-Plex Complexation Buffer
• 100 mL AAV-MAX Lysis Buffer
• 100 mL GlutaMAX Supplement

Storage conditions vary

Kidney (Embryonic)
AAV Production System
AAV Production
Sufficient for 1 L of Culture
Viral Production Cells 2.0
Suspension Cell Culture
Sterile
AAV-MAX
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.