Learn More
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Supplier: Thermo Scientific™ AB0564
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Highlights
- Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
- Each well has an independent sealing rim to prevent cross-contamination
- U-bottomed wells ideally suited for sample resuspension
- Polypropylene (AB-1068, polystyrene)
- Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
- Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
- Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
- Cleanroom manufactured
- ANSI-format for compatibility with automated systems
Specifications
Custom Barcoding Available | |
96-well | |
8 x 12 | |
Individually Wrapped | |
Abgene | |
96-well plate | |
Polypropylene | |
96 | |
Cleanroom | |
Round | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
1200 μL | |
1000 μL | |
Non-treated | |
Yes | |
2000 x g | |
1 | |
Yes | |
ANSI/ SLAS Standard |