missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate

Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.

Supplier:  Thermo Scientific™ AB0787

 View more versions of this product

Catalog No. AB-0787


Explore available promotions

Description

Description

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

Thermo Scientific 1.2 mL Storage Plates come with either round or square shapes, and they are made of polypropylene. These plates are ideal for suspension cell culture and are compatible with automated systems. Different sealing options are available for the 1.2 mL Storage Plates, and you can seal each well independently to prevent cross-contamination.
Highlights
• Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
• Each well has an independent sealing rim to prevent cross-contamination
• U-bottomed wells ideally suited for sample resuspension
• Polypropylene (AB-1068, polystyrene)
• Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
• Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
•Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
• Cleanroom manufactured
• ANSI-format for compatibility with automated systems
Applications
• Sample storage
• Sample resuspension
• Cell culture
• Assay setup: diluting and aliquoting

Compatible Sealing Film:Adhesive Heat
> Hazardous:No
> Max Centrifugal Force:2000 g (AB-0564 and AB-1127)
> Max Vol (Cap Seal):1.1 mL (AB-0564 only)
> Max Vol (Film Seal):1.2 mL
> Max Vol (Mat Seal):0.85 mL (AB-0564, AB-1068), 1 mL (AB-1127)
> Storage Condition:Ambient

Sample storage, sample resuspension, cell culture and assay setup: diluting and aliquoting.

Certifications

Free from human DNA, RNases, and DNases.

Specifications

Specifications

Custom Barcoding Available
96-well
8 x 12
Bulk
Abgene
96-well plate
Polypropylene
96
Cleanroom
Round
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
50
For Research Use Only. Not for use in diagnostic procedures.
PCR Ready
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
Natural
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
1200 μL
1000 μL
Non-treated
Yes
2000 x g
50
Yes
ANSI/ SLAS Standard
Videos
SDS
Documents

Documents

Promotions

Promotions