Learn More
Abietic acid, 85%, Thermo Scientific Chemicals
$150.54 - $341.91
Chemical Identifiers
CAS | 514-10-3 |
---|---|
Molecular Formula | C20H30O2 |
Molecular Weight (g/mol) | 302.46 |
MDL Number | MFCD03423567 |
InChI Key | RSWGJHLUYNHPMX-ONCXSQPRSA-N |
Synonym | abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 |
PubChem CID | 10569 |
ChEBI | CHEBI:28987 |
IUPAC Name | (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
SMILES | CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC173130250
|
Thermo Scientific Chemicals
173130250 |
25 g | Glass bottle |
Each for $150.54
|
|
||||
AC173131000
|
Thermo Scientific Chemicals
173131000 |
100 g | Glass bottle |
Each for $341.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
514-10-3 | |
302.46 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
10569 | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
C20H30O2 | |
MFCD03423567 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
CHEBI:28987 | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Specifications
514-10-3 | |
100.0 | |
85% | |
MFCD03423567 | |
15, 7 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
10569 | |
302.45 | |
Technical | |
Authentic | |
Crystals and Chunks |
80.0 | |
Brown-Yellow | |
C20H30O2 | |
09, II, 424 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 | |
302.46 | |
CHEBI:28987 | |
≥80% | |
25 g | |
Glass bottle | |
Abietic acid, 85% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 208-178-3
RTECSNumber : TP8580000
TSCA : TSCA
RUO – Research Use Only