Applied Biosystems™ Absolute Q™ 1-step RT-dPCR Master Mix (4X)
Applied Biosystems Absolute Q 1-step RT-dPCR Master Mix (4X) is optimized for use with the QuantStudio Absolute Q Digital PCR System in a simple workflow with minimal processing steps.
Supplier: Applied Biosystems™ A55146
Description
- Optimized for use with the QuantStudio Absolute Q Digital PCR System in a simple workflow with minimal processing steps
- The 4X formulation enables analysis of higher sample volumes and delivers accurate quantification of RNA targets without using a standard curve
- Set up the reaction like 1-Step RT-qPCR, then load and run the plate in a single instrument
- Requires just 5 minutes of hands-on time and RT-dPCR run time of ∼65 minutes
- Ability to detect less than 1 copy/μL of a specific target within a 4-plex reaction
- Detect up to four targets in a single reaction
- Highly reproducible results between technical replicates, consistently generate >20,000 analyzable micro-chambers
Specifications
Primer-probe | |
Taq DNA Polymerase | |
Digital PCR | |
Absolute Q Digital PCR System | |
200 Reactions | |
450 μL |
Digital PCR | |
<2 hr. | |
200 reactions | |
Hot Start Taq Polymerase | |
RNA |
For research use only. Not for use in diagnostic procedures.