Learn More
Applied Biosystems™ Absolute Q™ Liquid Biopsy dPCR Assay - KIT 1314
Absolute Q™ Liquid Biopsy dPCR assays enable reproducible, specific detection of known somatic mutations.
Supplier: Applied Biosystems™ A52757
Description
Each predesigned Absolute Q™ Liquid Biopsy dPCR assay has been validated using controls to detect down to 0.1% variant allele frequency in genes relevant for cancer research.Spécifications
| 40X | |
| Tube | |
| Absolute Q | |
| Assay |
| Digital PCR | |
| Digital PCR | |
| QuantStudio™ Absolute Q™ Digital PCR System | |
| 700 |