missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Absorption Cylindrical Cells
Fit standard spectrophotometer cell holders
Supplier: Fisherbrand™ 14958143
Includes: A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
Description
Fit standard spectrophotometer cell holders
- All cells are 22mm wide
- Available size is macro
- 10, 20, 50 or 100mm path length rectangular cells
- Available with a variety of window material for wide range of assays
- Available in two materials e.g. Quartz and Special Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Cell | |
Quartz | |
100 mm | |
35 mm | |
Cylinder |
31,000 μL | |
18.5mm | |
190 to 2600 nm and 2850 to 3600 nm | |
22 mm |