missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acarbose, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 459080010

 View more versions of this product

Catalog No. AC459080010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acarbose
90.0°C
Authentic
Glass Bottle
1 g
acarbose
XUFXOAAUWZOOIT-AKWDJIIVSA-N
(3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
131674886
95%
56180-94-0
5% max.
94% min. (HPLC)
C25H43NO18
+168° to +183° (20°C, 589nm) (1% in water)
Solubility in water: soluble. Other solubilities: slightly soluble in ethanol
CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO
645.61
645.61

Chemical Identifiers

56180-94-0
645.61
acarbose
(3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
C25H43NO18
XUFXOAAUWZOOIT-AKWDJIIVSA-N
131674886
CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 260-030-7

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only