Learn More
Thermo Scientific™ AccelerOme™ Label-Free MS Sample Prep Kit
Kit provides ready-to-go reagents for efficient and robust proteomics mass spectrometry (MS) sample preparation for use with the AccelerOme Automated Sample Prep System.
$1379.95 - $2207.94
Specifications
| Product Type | Reagent Kit |
|---|---|
| Product Line | Thermo Scientific |
| Starting Material | Protein Samples |
| Detection Method | UV Absorption |
Description
- Kit contains pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean up cartridges for the processing of samples from protein lysis and digestion through peptide clean-up
- The single-use components are conveniently provided in labeled trays for direct placement on the AccelerOme system
- A peptide retention time calibration standard is also included with the kit (shipped separately) that can be spiked into the sample before LC-MS analysis to assess system performance.
- Complete set of ready-to-use proteomic sample prep and clean-up reagents for easy loading onto instrument
- Reagents combined with AccelerOme ecosystem provide consistent, high-quality results with technical replicates with <5% CV
- Reagents verified on system using cells, plasma, proteins, and tissue samples for 1–100 μg samples
- Instrumentation and reagents perform the work enabling time away from tedious hands-on sample preparation
Specifications
| Reagent Kit | |
| Thermo Scientific | |
| Protein Samples | |
| UV Absorption |