missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Access RT-PCR System DFS Item

Access RT-PCR System is designed for the reverse transcription and PCR amplification of a specific target RNA from total RNA or mRNA.

Supplier:  Promega A1260

 View more versions of this product

Catalog No. PR-A1260


Explore available promotions

Description

Description

  • Convenient RT-PCR in One Tube
  • All the reagents you need for single-tube reverse transcription and PCR
  • Separate components allow you to control every step of the reaction
Specifications

Specifications

Contains sufficient reagents for 20 reactions
• Positive Control RNA and Primers
• AMV Reverse Transcriptase, 100 μL
• Tfl DNA Polymerase, 100 μL
• AMV/Tfl 5X reaction Buffer, 1 mL
• MgSO, 1.25 mL
• dNTP Mixture, 20 μL
• 50 μL Positive Control

Store at –30° to –10°C

20 Reactions
20 Reactions
Access RT-PCR introductory system
RT-PCR
Videos
SDS
Documents

Documents

Promotions

Promotions