Thermo Scientific™ Accessories for Digital Heating Cooling Drybath
Optimize the performance of our digital heating cooling drybath and digital heating shaking bath by using these accessories.
$739.85 - $1289.38
Catalog Number | Mfr. No. | No. of Places | Tube Size | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Places | Tube Size | Type | Price | Quantity | |||
11686738
|
Thermo Scientific™
88880129 |
48 | 0.5mL | Block |
Each for $787.96
|
|
|||
11686739
|
Thermo Scientific™
88880130 |
30 | 1.5mL | Block |
Each for $739.85
|
|
|||
11686740
|
Thermo Scientific™
88880131 |
15 | 15mL | Block |
Each for $800.40
|
|
|||
11686741
|
Thermo Scientific™
88880132 |
6 | 50mL small | Block |
Each for $956.13
|
|
|||
11686742
|
Thermo Scientific™
88880133 |
6 | 50mL large | Block |
Each for $1,289.38
|
|
|||
11686743
|
Thermo Scientific™
88880134 |
35 | 10mm | Block |
Each for $787.96
|
|
|||
11686744
|
Thermo Scientific™
88880135 |
24 | 12mm | Block |
Each for $787.96
|
|
|||
11686745
|
Thermo Scientific™
88880136 |
24 | 13mm | Block |
Each for $787.96
|
|
|||
11686746
|
Thermo Scientific™
88880137 |
20 | 15mm | Block |
Each for $787.96
|
|
|||
11686747
|
Thermo Scientific™
88880138 |
16 | 16mm | Block |
Each for $833.66
|
|
|||
11686748
|
Thermo Scientific™
88880139 |
12 | 18mm | Block |
Each for $787.96
|
|
|||
11686749
|
Thermo Scientific™
88880140 |
12 | 20mm | Block |
Each for $787.96
|
|
|||
Description
Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.
- Warranty: 2 years
Specifications
LED | |
Block |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.