Labconco™ Accessories for Labconco™ Kjeldahl Rapid Digester-25
Required for use with Labconco Rapid Digestors
Supplier:  Labconco™ 2304005
 View more versions of this product
Catalog No. 13-162-11
Explore available promotions
Description
Description
  • Designed for uniform wall thickness and impact strength
  • Made of borosilicate glass with heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock
  • Volumetric tubes have an easy-to-read calibration mark for accurate dilutions and a built-in constriction for maximum refluxing and digestion speed
Specifications
Specifications
Straight Digestion Tube
250 mL
Borosilicate Glass
8.45 oz.
Kjeldahl rapid digestor
5 Pk.