Sper Scientific™ Accessories for Sper Scientific Meters
Intelligent probes electronically retain calibration information.
Supplier: Sper Scientific™ 840016
Description
- Uses interchangeable probes to read pH, mV, conductivity, TDS, DO temperature and ORP
Specifications
BNC connector | |
±(0.02 pH + 2d) | |
Water quality testing | |
15.2cm |
6 in. | |
pH Probe | |
Sper Scientific meters |