Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
The touch-screen reader has 8GB on board memory to provide virtually unlimited data logging capacity
$34.46 - $51.69
Specifications
For Use With (Equipment) Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40)
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
13-245-302
View Documents
Digi-Sense™
2025047
Extension Sensor Cable
Each for $51.69
13-245-305
View Documents
Digi-Sense™
2025049
Replacement USB Cable
Each for $34.46
Description
Description
  • Includes A2LA accredited NIST-Traceable Calibration Certificate
  • Measures, logs, and displays temperature, humidity, and dew point
  • Removable sensor for local or remote measurements using extension cable included
  • Wall mount or desk mount operation
  • Resident self-booting software, no installation necessary
  • Comes with NIST calibration
  • Stored data downloads to computer via USB cable provided in CSV format
Warranty
2 Years
Specifications
Specifications
Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'