BD Accessories for Veritor™ Plus Analyzer
Accessories for use with Veritor Plus Analyzer.
$22.57 - $526.54
Specifications
Description For use with Veritor Plus Analyzer
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
B256068
View Documents
BD
256068
InfoScan Module
Each for $526.54
B443907
View Documents
BD
443907
Printer Cable
Each for $22.57
Description
Description
InfoScan Module
  • Point of Care scanning of three different identifiers - Operator ID, Specimen ID, and Test Kit/Lot information
  • Identifiers can be locally recorded, printed or downloaded and are automatically documented
  • Barcode scanner
  • Facilitate traceability and help reduce manual transcription
  • Data access in a CSV file downloaded via USB to a local computer or print-enabled
Printer Cable
  • Custom printer cable to connect to Epson TM-M30, TM-L90 or TM-M20II
Specifications
Specifications
For use with Veritor Plus Analyzer
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'