missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bioscience accu-Test™ Vials for COD Testing
For low-, standard- and high-range concentrations
$349.33 - $524.66
Specifications
Certifications/Compliance | EPA, OSHA |
---|---|
For Use With (Equipment) | Standard COD reactor and spectrophotometer |
Includes | 16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap |
Catalog Number | Mfr. No. | Composition | For Use With (Application) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Composition | For Use With (Application) | Price | Quantity | ||||
01-215-28
|
Bioscience
185252 |
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid | COD testing for 20 to 900mg/L standard range concentration |
Pack of 50 Each for $349.33
Case of 3 Pack for $1,047.99
|
|
||||
01-215-22B
|
Bioscience
174334 |
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid, 0.60% Mercuric Sulfate | COD testing for 5 to150mg/L standard range concentration |
Pack of 50 Each for $524.66
|
|
||||
01-215-22C
|
Bioscience
174326 |
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid, 0.60% Mercuric Sulfate | COD testing for 100 to 4500mg/L standard range concentration |
Pack of 50 Each for $524.66
|
|
||||
01-215-22A
|
Bioscience
174318 |
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid, 0.60% Mercuric Sulfate | COD testing for 20 to 900mg/L standard range concentration |
Pack of 50 Each for $524.66
|
|
||||
Description
- Convenient to use
- Give excellent analytical results
- Compatible with all standard COD reactors and spectrophotometers
- For samples with low chloride and ammonia levels, mercury-free reagents are available for nonreporting purposes
Order Info
Supplied in modular packs that are easy to stack
Certifications
U.S. EPA approved (for standard and low ranges) micro Chemical Oxygen Demand test
Specifications
EPA, OSHA | |
Standard COD reactor and spectrophotometer | |
16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap |