missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Hazardous Waste Labels
Ensure the contents in containers are clearly identified and proper precautions are addressed with Container Identification Labels
Supplier: Accuform Signs MHZW09EVP
Spécifications
| Self-adhesive | |
| Vinyl | |
| Hazardous Waste Federal/ State Law | |
| 25/Pk. | |
| 15.2 x 15.2 cm | |
| -20°C to +80°C |
| Label | |
| Hazardous Waste Label | |
| Hazardous waste | |
| 6 x 6 in. | |
| -20 to 176°F |