missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Accuform Signs NFPA Classification Roll Labels

to ensure the quick identification of fire hazards caused by chemicals

Supplier:  Accuform Signs LZN646PS5

 View more versions of this product

Catalog No. 19-121-465


Explore available promotions

Description

Description

Specifications

Specifications

Polyester
6 x 6 in.
Self-adhesive
-40°C to +121°C
NFPA Label
500/Pk.
15.24 cm
Hazard communication
-40°F to +250°F
White
Videos
SDS
Documents

Documents

Promotions

Promotions