missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs NFPA Diamond Roll Labels
Quickly identify any serious hazards present associated with the chemicals.
$56.22 - $280.32
Specifications
| Mounting Type | Self-adhesive |
|---|---|
| For Use With (Application) | Hand-written hazard codes and symbols |
| Product Type | NFPA Label |
| Color | White |
| Catalog Number | Mfr. No. | Material | Size (English) | Quantity | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Size (English) | Quantity | Price | Quantity | |||
|
19-121-455
|
Accuform Signs
LZN631EV2 |
Polyester | 2 x 2 in. | 250/Pk. |
Pack of 250 Each for $89.74
|
|
|||
|
19-121-457
|
Accuform Signs
LZN633EV2 |
Polyester | 4 x 4 in. | 250/Pk. |
Pack of 250 Each for $131.42
|
|
|||
|
19-121-456
|
Accuform Signs
LZN633PS2 |
Paper | 4 x 4 in. | 250/Pk. |
Pack of 250 Each for $101.06
|
|
|||
|
19-121-459
|
Accuform Signs
LZN635EV2 |
Polyester | 6 x 6 in. | 250/Pk. |
Pack of 250 Each for $175.46
|
|
|||
|
19-121-458
|
Accuform Signs
LZN635PS2 |
Paper | 6 x 6 in. | 250/Pk. |
Pack of 250 Each for $126.41
|
|
|||
|
19-804-614
|
Accuform Signs
LZN633PS5 |
Paper | 4 x 4 in. | 500/Pk. |
Each for $161.62
|
|
|||
|
19-804-611
|
Accuform Signs
LZN631EV5 |
Polyester | 2 x 2 in. | 500/Pk. |
Each for $126.02
|
|
|||
|
19-804-613
|
Accuform Signs
LZN631PS5 |
Paper | 2 x 2 in. | 500/Pk. |
Each for $90.78
|
|
|||
|
19-804-615
|
Accuform Signs
LZN635EV5 |
Polyester | 6 x 6 in. | 500/Pk. |
Each for $280.32
|
|
|||
|
19-804-616
|
Accuform Signs
LZN633EV5 |
Polyester | 4 x 4 in. | 500/Pk. |
Each for $224.33
|
|
|||
|
19-121-454
|
Accuform Signs
LZN631PS2 |
Paper | 2 x 2 in. | 250/Pk. |
Pack of 250 Each for $56.22
|
|
|||
Specifications
| Self-adhesive | |
| Hand-written hazard codes and symbols | |
| NFPA Label | |
| White |