Accuform Signs Pipe-Marking Tape
Solid color marking tapes increase pipe visibility and help to identify hazardous areas.
Supplier:  Accuform Signs PTM613RD
 View more versions of this product
Catalog No. 19-805-168
Explore available promotions
Description
Description
Specifications
Specifications
OSHA 1910.144
Red
1 Roll
-20 to 140°F
6 mil
32.92 m
Self-adhesive
Vinyl
Pipe Marker
-28.88 to 60°C
108 ft.
1 in.