missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Accuform Signs Solid Color Marking Tapes
High-visibility marking of hazard areas
Supplier: Accuform Signs PTM626OR
Description
- Size (W × L): 2.5cm × 33m (1 in. × 108.3 ft.)
- 6-mil vinyl tape
- Waterproof
- Wear-resistant
- Minor resistance to acids, oils, grease, caustics, alkalis and most solvents
- Aggresive, permanent, self-adhesive back applies to clean, smooth surface
Color
- Red: For fire protection equipment and danger hazards
- Orange: For danger points on machines
Specifications
1 Roll | |
Self-adhesive | |
Vinyl | |
180 ft. x 2 in. (54.9 m x 2.5 cm) | |
2 in. | |
Orange | |
54.9 m |
180 ft. | |
Used for color-coding and identifying aisles, walkways, indoor traffic lines, danger zones, boundaries, doorways and any other general purpose marking | |
-28.89°C to +80°C | |
6 mil | |
-20°F to +176°F | |
2.5 cm | |
OSHA 1910.144 |