missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ AB200 Benchtop pH/Conductivity Meters

Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB200 pH/conductivity meter combines the benefits of a pH meter and conductivity meter in one easy to use, reliable measurement system. The large backlit display offers great visibility in laboratory environments.

Supplier:  Fisherbrand™ ORI13636AB200

 View more versions of this product

Catalog No. 13-636-AB200


Explore available promotions

Includes: accumet AB200 pH benchtop meter, TRIS-compatible double junction refillable epoxy-body pH/ATC electrode (13-620-631), conductivity/temperature probe (13-620-100), electrode stand, RS232 and USB cables, 100/240V universal power supply

Description

Description

Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB200 pH/conductivity meter combines the benefits of a pH meter and conductivity meter in one easy to use, reliable measurement system. The large backlit display offers great visibility in laboratory environments.

  • Graphic backlit display with date and time, stability indicator and alarms
  • Dual channel meter for pH and conductivity measurements
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
NIST-Traceable Calibration Model (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate
Specifications
pH Range: -2.000 to 20.000 pH
pH Resolution: 0.1, 0.01, 0.001 pH
pH Accuracy: ±0.002 pH
mV Range: ±2000.0mV
mV Resolution: 0.1mV
mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
mV Offset Adjustment: Up to ±150mV
Conductivity Range: 0.00μS to 500.0mS
Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
Conductivity Accuracy: ±1 % full scale
TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
TDS Accuracy: ±1 % full scale
Salinity Range: 0 to 80.0 ppt
Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
Salinity Accuracy: ±1 % full scale
Resistivity Range: 2.000 Ohm to 20.0 MOhm
Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
Resistivity Accuracy: ±1 % full scale
Temperature Range: 0° to 100°C (32° to 212.0°F)
Temperature Resolution: 0.1°C (0.1°F)
Temperature Accuracy: ±0.3°C (±0.5°F)
Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
Self-Test: Yes
Inputs: BNC, mini-phone ATC, 8 Pin DIN, ground
Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Certifications

CE, TUV 3-1, FCC Class A

Warranty

3 years on meter

Specifications

Specifications

RS232 (phono plug), mini-B USB, stirrer
±0.2 mV
± 1% full scale
± 1% full scale
Up to 6 preset or 5 custom
Without NIST-traceable pH Certificate
500
LCD
0 μS to 500 mS
-2 to 20
0 to 80 ppt
0.1/0.01/0.001
Meter, TRIS Compatible pH/ATC Electrode (13-620-631), Conductivity/Temp Probe (13-620-100), Electrode Arm, RS-232 and USB cables, 100/240V Power Supply
AB200 pH/Conductivity Kit
0.1mV
0.01/0.1ppm, 0.001/0.01/0.1ppt
±0.3°C
3 Years
50/60Hz
±1% FS
±0.002
± 1% full scale
Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
0.010 to 10.000
Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format
2
pH, mV, Conductivity, TDS, Salinity, Resistivity, Temperature
±2000 mV
2 W to 20 MW
0 ppm to 500 ppt (at TDS factor 1)
0°C to 100°C
BNC, ATC
0.01/0.1μS, 0.001/0.01/0.1mS
0.01/0.1W, 0.001/0.1kW, 0.01MW
0/0.1ppm, 001/01/0.1ppt
0.1°C
100/240V
SDS
Documents

Documents

Promotions

Promotions