missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

accumet™ Standard All-in-One pH Buffer Kit, Fisherbrand™

Supplier:  Fisherbrand ORI910199FS

Catalog No. 15200909


Explore available promotions

Description

Description

Specifications

Specifications

pH Buffer, storage solutions
Accurate calibrations and NIST traceability
4 x 475 mL
Bottle
Standard All-in-One pH Buffer Kit
pH Meter and Electrode
Buffer, Storage
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions