Fisherbrand™ accuSpin™ Micro 21/21R Microcentrifuges
The Fisherbrand™ accuSpin™ Micro 21/21R Microcentrifuge with 24 x 1.5/2.0 mL rotor with ClickSeal™ biocontainment lid offers exceptional acceleration of up to 21,000 xg in as little as 12 seconds.
Supplier:  Fisherbrand™ 75003247
 View more versions of this product
Catalog No. 13100678
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: accuSpin Micro 21R Microcentrifuge, 24 x 1.5/2 mL rotor with ClickSeal™ biocontainment lid
Description
Description
The accuSpin Micro 21R is refrigerated.
  • Durable and flexible rotors
  • Outstanding corrosion resistance with lightweight engineered polymer rotors
  • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
  • ClickSeal™ Biocontainment solution
  • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with ClickSeal™ biocontainment lids
  • Quiet, Easy to Use Design
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
  • Sleek, lightweight, space-saving design
  • Durable and easy to clean and maintain
  • Quiet performance promotes a safe, stress-free environment
Specifications
Specifications
24 x 1.5/2 mL
21,000 x g
24 x 1.5/2 mL rotor with ClickSeal™ biocontainment lid
No
Yes
Yes
28 kg (62 lb.)
14,800 rpm
FDA listed, Product code JQC centrifuges for clinical use, Device class 1, ANSI/UL 61010-1 3.1 Edition, UL 61010-2-020 3rd Edition, UL 61010-2-011 2nd Edition, UL 61010-2-101 3rd Edition, FCC Part 15, ICES-001, EN ISO 14971, ISO 13485
4.4 A
accuSpin Micro 21R (Refrigerated)
2 years parts and labor
33.0 x 29.5 x 44.5 cm (12.9 x 11.6 x 17.5 in.)
Fisherbrand