Fisherbrand™ accuSpin™ 8C Small Benchtop Centrifuge
Perform both routine clinical and life science separations with this compact centrifuge and the unique DualSpin™ rotor featuring a 2-in-1 hybrid design
Supplier:  Fisherbrand™ 75008821
Catalog No. 75-008-821
Explore available promotions
Description
Description
Innovative DualSpin rotor
  • Unique “2-in-1” hybrid design with interchangeable fixed angle and swinging buckets
  • No need to change rotor
  • Able to accommodate 1.4 mL to 15 mL common blood tubes, geltubes, 10 mL standard syringes and 15 mL conical tubes
Versatile functionality
  • Large, brightly-lit display is easy to read and fast one-click lid closure simplifies operation
  • Flexible setting of running parameters
  • 4 program memory
  • 3 selections of deceleration profiles, including brake-off
User-friendly design
  • Light weight composite rotor with good chemical compatibility
  • Quiet operation at noise level less than 56 dBA
  • Fast one-click centrifuge lid closure
  • Large, brightly lit display with intuitive controls
  • Conforms to the latest clinical and safety standards
Specifications
Specifications
DualSpin rotor body, fixed angle buckets (set of 8), swinging buckets (set of 8), green/short spacers (set of 8), yellow/long spacers (set of 8)
3,114 x g (fixed angle); 3,490 x g (swinging bucket)
No
120 V 60 Hz
Large, Bright LED
240 mm
450 mm
12.79 in.
130 w
17.68 x 12.77 x 17.68 in. (240 x 325 x 450 mm)
20.08 in.
8 x 15 mL, 8 x 10 mL
4,900 rpm
<56 dB(A) (swinging bucket)
UL, CE, RoHS, CSA, IVD, FDA class 1
9.45 in.
17.79 in.
Lid lock, steel armored ring, view port for speed calibration, lid emergency release
325 mm
Microprocessor
1-99 minutes, in increments of 1 minute; HOLD
510 mm