missing translation for 'onlineSavingsMsg'
Learn More

Acemetacin

A potent inhibitor of COX-2 found to have anti-tumor activity in the colon | CAS: 53164-05-9 | C21H18ClNO6 | 415.826 g/mol

Supplier:  Thermo Scientific Chemicals J6358304

 View more versions of this product

Catalog No. AAJ6358304

Only null left
Explore available promotions

Description

Description

A potent inhibitor of COX-2 found to have anti-tumor activity in the colon

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acemetacin
149°C to 153°C
C21H18ClNO6
2 g
14,27
FSQKKOOTNAMONP-UHFFFAOYSA-N
2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid
1981
415.83
53164-05-9
Yellow
MFCD00151473
UN2811
acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O
415.826
CHEBI:31162
Powder

Chemical Identifiers

53164-05-9
415.826
FSQKKOOTNAMONP-UHFFFAOYSA-N
1981
2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid
C21H18ClNO6
MFCD00151473
acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester
CHEBI:31162
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c

H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 258-403-4

RTECSNumber : NL3521400

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only