Learn More
Acetal, 99%, Thermo Scientific Chemicals
$52.85 - $387.49
Chemical Identifiers
CAS | 105-57-7 |
---|---|
Molecular Formula | C6H14O2 |
Molecular Weight (g/mol) | 118.18 |
MDL Number | MFCD00009243 |
InChI Key | DHKHKXVYLBGOIT-UHFFFAOYSA-N |
Synonym | acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano |
PubChem CID | 7765 |
IUPAC Name | 1,1-diethoxyethane |
SMILES | CCOC(C)OCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC102220050
|
Thermo Scientific Chemicals
102220050 |
5 mL | Glass bottle |
N/A
|
|
||||
AC102222500
|
Thermo Scientific Chemicals
102222500 |
250 mL | Glass bottle |
N/A
|
|
||||
AC102220010
|
Thermo Scientific Chemicals
102220010 |
1 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-57-7 | |
118.18 | |
DHKHKXVYLBGOIT-UHFFFAOYSA-N | |
7765 | |
CCOC(C)OCC |
C6H14O2 | |
MFCD00009243 | |
acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano | |
1,1-diethoxyethane |
Specifications
105-57-7 | |
100.0 | |
Colorless | |
103.0°C | |
Authentic | |
Glass bottle | |
1.3790 to 1.3850 | |
MFCD00009243 | |
01, 603 | |
15, 39 | |
Solubility in water: 46g/L (25°C). Other solubilities: miscible with alcohol,ether,soluble in heptane,methylcyclohexane,,ethyl acetate,propyl-,isopropyl-,butyl-,,isobutyl alcohol | |
CCOC(C)OCC | |
118.18 | |
118.18 | |
Pure | |
Acetal, 99% |
98.5 | |
-100.0°C | |
0.8250g/mL | |
−21°C | |
98.5% min. (GC) | |
C6H14O2 | |
CH3CH(OC2H5)2 | |
5 mL | |
0.825 | |
acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano | |
DHKHKXVYLBGOIT-UHFFFAOYSA-N | |
1,1-diethoxyethane | |
7765 | |
99% | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-310-6
RTECSNumber : AB2800000
TSCA : TSCA
RUO – Research Use Only