missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetaldehyde, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals

$122.74 - $122.74

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.04
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427171000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
427171000
100 mL AcroSeal™ Glass bottle
Each for $122.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-07-0
44.04
IKHGUXGNUITLKF-UHFFFAOYSA-N
177
acetaldehyde
C2H4O
MFCD00006991
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
CHEBI:15343
CC=O

Specifications

75-07-0
100.0
White
21.0°C
Authentic
AcroSeal™ Glass bottle
CH3CHO
100 mL
15,4
Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone
CC=O
44.04
CHEBI:15343
99.5%
Solid
99.4
-123.0°C
0.7850g/mL
−27°C
99.4% min. (GC)
C2H4O
MFCD00006991
0.785
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
177
44.04
Extra Pure
Acetaldehyde, Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger

EINECSNumber : 200-836-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only