Learn More
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
$62.52 - $175.26
Chemical Identifiers
CAS | 58052-80-5 |
---|---|
Molecular Formula | C6H18N3 |
Molecular Weight (g/mol) | 132.23 |
MDL Number | MFCD00149559 |
InChI Key | MZSSRMMSFLVKPK-UHFFFAOYSA-Q |
Synonym | 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt |
PubChem CID | 2723814 |
SMILES | CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC148530250
|
Thermo Scientific Chemicals
148530250 |
25 g |
Each for $62.52
|
|
|||||
AC148531000
|
Thermo Scientific Chemicals
148531000 |
100 g |
Each for $175.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58052-80-5 | |
132.23 | |
MZSSRMMSFLVKPK-UHFFFAOYSA-Q | |
2723814 |
C6H18N3 | |
MFCD00149559 | |
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt | |
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 |
Specifications
58052-80-5 | |
100.0 | |
Beige | |
55°C | |
98% | |
C6H18N3 | |
25 g | |
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt | |
MZSSRMMSFLVKPK-UHFFFAOYSA-Q | |
2,4,6-trimethyl-1,3,5-triazinane;trihydrate | |
2723814 | |
98% | |
Acetaldehyde ammonia trimer, 98% |
97.5 | |
95.0°C to 97.0°C | |
110.0°C | |
Authentic | |
Glass bottle | |
MFCD00149559 | |
26, I, 3 | |
Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform | |
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 | |
132.23 | |
183.26 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
EINECSNumber : 200-868-2
RUO – Research Use Only