missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetaldehyde dimethyl acetal, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1093922

 View more versions of this product

Catalog No. AAA1093922


Explore available promotions

Description

Description

Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Acetaldehyde dimethyl acetal
-113°C
63°C to 65°C
Sweet
1.367
100 g
1697039
14,3226
Soluble in water (1000mg/ml at 25°C),alcohols,chloroform,ether,and acetone.
CC(OC)OC
90.122
90.12
534-15-6
0.852
−17°C (1°F)
C4H10O2
MFCD00008493
UN2377
Moisture sensitive
dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane
SPEUIVXLLWOEMJ-UHFFFAOYSA-N
1,1-dimethoxyethane
10795
≥98%

Chemical Identifiers

534-15-6
90.122
SPEUIVXLLWOEMJ-UHFFFAOYSA-N
10795
CC(OC)OC
C4H10O2
MFCD00008493
dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane
1,1-dimethoxyethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,1-DIMETHOXYETHANE

EINECSNumber : 208-589-8

RTECSNumber : AB2825000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only