missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetaldehyde ethyl propargyl acetal, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 390640050

 View more versions of this product

Catalog No. AC390640050


Explore available promotions

Description

Description

Specifications

Specifications

Acetaldehyde ethyl propargyl acetal
97.5
Colorless to Yellow
141°C to 142°C
97.5% min. (GC)
C7H12O2
MFCD00066344
0.893
QKBBTQJLUGADEG-UHFFFAOYSA-N
3-(1-ethoxyethoxy)prop-1-yne
86774
98%
18669-04-0
100.0
0.8930g/mL
Authentic
Glass bottle
1.4150 to 1.417
5 g
3-1-ethoxyethoxy prop-1-yne, acetaldehyde ethyl propargyl acetal, 1-propyne, 3-1-ethoxyethoxy, 1-ethoxy-1-prop-2-yn-1-yloxy ethane, 3-1-ethoxyethoxy propyne, 1-ethoxyethoxy-2-propyne, 1-1-ethoxyethoxy-2propyne, 1-1-ethoxyethoxy-2-propyne, 5-methyl-4,6-dioxa-1-octyne, 1-propyne,3-1-ethoxyethoxy
CCOC(C)OCC#C
128.17
128.17
Liquid

Chemical Identifiers

18669-04-0
128.17
QKBBTQJLUGADEG-UHFFFAOYSA-N
3-(1-ethoxyethoxy)prop-1-yne
C7H12O2
MFCD00066344
86774
CCOC(C)OCC#C
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-490-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only