missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetamide, 99%, pure
CAS: 60-35-5 | C2H5NO | 59.06 g/mol
$79.71 - $173.33
Chemical Identifiers
| CAS | 60-35-5 |
|---|---|
| Molecular Formula | C2H5NO |
| Molecular Weight (g/mol) | 59.06 |
| InChI Key | DLFVBJFMPXGRIB-UHFFFAOYSA-N |
| Synonym | ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van |
| PubChem CID | 178 |
| ChEBI | CHEBI:49028 |
| IUPAC Name | acetamide |
| SMILES | CC(=O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC153631000
|
Thermo Scientific Chemicals
153631000 |
100 g | Plastic bottle |
Each for $79.71
|
|
||||
|
AC153635000
|
Thermo Scientific Chemicals
153635000 |
500 g | Plastic bottle |
Each for $173.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 60-35-5 | |
| 59.06 | |
| ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van | |
| CHEBI:49028 | |
| CC(=O)N |
| C2H5NO | |
| DLFVBJFMPXGRIB-UHFFFAOYSA-N | |
| 178 | |
| acetamide |
Specifications
| 60-35-5 | |
| White | |
| Authentic | |
| Plastic bottle | |
| CH3CONH2 | |
| 02,II,177 | |
| 15,44 | |
| Solubility in water: 2000g/L (20°C). Other solubilities: 1g/2 mL alcohol- 1g/6 mL pyridine,soluble in chloroform,glycerol and hot benzene,slightly soluble in ether | |
| CC(=O)N | |
| 59.06 | |
| CHEBI:49028 | |
| 99% | |
| Crystals |
| 79.0°C to 81.0°C | |
| 221.0°C | |
| 98.5% min. (GC) | |
| C2H5NO | |
| 100 g | |
| 01,3 | |
| ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van | |
| DLFVBJFMPXGRIB-UHFFFAOYSA-N | |
| acetamide | |
| 178 | |
| 59.06 | |
| Pure | |
| Acetamide |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 200-473-5
RUO – Research Use Only