missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetamidine hydrochloride, 97%, Thermo Scientific Chemicals

$49.48 - $425.49

Chemical Identifiers

CAS 124-42-5
Molecular Formula C2H7ClN2
Molecular Weight (g/mol) 94.542
MDL Number MFCD00013016
InChI Key WCQOBLXWLRDEQA-UHFFFAOYSA-N
Synonym acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
PubChem CID 67170
IUPAC Name ethanimidamide;hydrochloride
SMILES CC(=N)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1609114
View Documents
Thermo Scientific Chemicals
A1609114
25 g
Each for $49.48
 
AAA1609122
View Documents
Thermo Scientific Chemicals
A1609122
100 g
Each for $120.90
 
AAA1609136
View Documents
Thermo Scientific Chemicals
A1609136
500 g
Each for $425.49
 
Description

Description

Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

Solubility
Soluble in water.

Notes
It is hygroscopic. Store away from oxidizing agents.
Specifications

Chemical Identifiers

124-42-5
94.542
WCQOBLXWLRDEQA-UHFFFAOYSA-N
67170
CC(=N)N.Cl
C2H7ClN2
MFCD00013016
acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
ethanimidamide;hydrochloride

Specifications

124-42-5
C2H7ClN2
25 g
Hygroscopic
acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
WCQOBLXWLRDEQA-UHFFFAOYSA-N
ethanimidamide;hydrochloride
67170
97%
168°C to 175°C
MFCD00013016
3591762
14,44
Soluble in water.
CC(=N)N.Cl
94.542
94.54
Acetamidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-700-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only