missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, 98%

CAS: 103-84-4 | C8H9NO | 135.17 g/mol

$68.68 - $448.38

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
Synonym acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
PubChem CID 904
ChEBI CHEBI:28884
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1436122
View Documents
Thermo Scientific Chemicals
A1436122
100 g
Each for $68.68
Only null left
 
AAA1436136
View Documents
Thermo Scientific Chemicals
A1436136
500 g
Each for $132.92
Only null left
 
AAA143610E
View Documents
Thermo Scientific Chemicals
A143610E
2500 g
Each for $448.38
Only null left
 
Description

Description

Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

Solubility
Soluble in ethanol, diethyl ether, acetone, benzene. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

103-84-4
135.17
FZERHIULMFGESH-UHFFFAOYSA-N
904
N-phenylacetamide
C8H9NO
MFCD00008674
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
CHEBI:28884
CC(=O)NC1=CC=CC=C1

Specifications

103-84-4
1.121
161°C (321°F)
C8H9NO
100 g
14,50
Soluble in ethanol,diethyl ether,acetone,benzene. Slightly soluble in water.
CC(=O)NC1=CC=CC=C1
135.17
CHEBI:28884
98%
113°C to 116°C
304°C to 305°C
98%
MFCD00008674
606468
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
FZERHIULMFGESH-UHFFFAOYSA-N
N-phenylacetamide
904
135.17
Acetanilide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 203-150-7

RTECSNumber : AD7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only