missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetanilide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150810010

 View more versions of this product

Catalog No. AC150810010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetanilide
99.0
111°C to 115°C
4.65 (Air = 1.0)
173°C
Conforms to Reference
Plastic bottle
CH3CONHC6H5
1 kg
15, 49
Solubility in water: slightly soluble (5g/L at 25°C). Other solubilities: 1g/20mL boiling water,1g/3.4mL alcohol,1g/4mL ac,etone,1g/3.7mL chloroform,1g/5mL glycerol,1g/8mL,dioxane,1g/18mL diethyl ether,1g/47mL benzene,,sparingly soluble in pe
CC(=O)NC1=CC=CC=C1
135.17
CHEBI:28884
13hPa at 114°C
Powder
103-84-4
100.0
Blue-Gray to White
304°C
≥111°C to ≤115°C
99% min. (GC)
C8H9NO
MFCD00008674
12, 237
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
FZERHIULMFGESH-UHFFFAOYSA-N
N-phenylacetamide
904
135.17
99+%

Chemical Identifiers

103-84-4
135.17
FZERHIULMFGESH-UHFFFAOYSA-N
904
N-phenylacetamide
C8H9NO
MFCD00008674
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
CHEBI:28884
CC(=O)NC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 203-150-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only