missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetazolamide, 99%, Thermo Scientific Chemicals

$69.40 - $167.64

Chemical Identifiers

CAS 59-66-5
Molecular Formula C4H6N4O3S2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00003105
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
Synonym acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
PubChem CID 1986
ChEBI CHEBI:27690
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0756206
View Documents
Thermo Scientific Chemicals
L0756206
5 g
Each for $69.40
 
AAL0756214
View Documents
Thermo Scientific Chemicals
L0756214
25 g
Each for $167.64
 
Description

Description

Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
Specifications

Chemical Identifiers

59-66-5
222.24
BZKPWHYZMXOIDC-UHFFFAOYSA-N
1986
N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
C4H6N4O3S2
MFCD00003105
acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
CHEBI:27690
CC(=O)NC1=NN=C(S1)S(N)(=O)=O

Specifications

59-66-5
C4H6N4O3S2
5 g
14,53
Soluble in NH4OH (50mg/mL),DMSO,Methanol and slightly soluble in Ethanol.
CC(=O)NC1=NN=C(S1)S(N)(=O)=O
222.24
CHEBI:27690
99%
∽255°C (decomposition)
MFCD00003105
212994
acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
BZKPWHYZMXOIDC-UHFFFAOYSA-N
N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
1986
222.25
Acetazolamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360

EINECSNumber : 200-440-5

RTECSNumber : AC8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only