missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid (100%, Glacial), Anhydrous, EMSURE™ MilliporeSigma™
$33.09 - $217.87
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1000632500
|
Technidata
1000632500 |
2.5 L | Glass bottle |
Each for $33.09
|
|
||||
M1000632510
|
Technidata
1000632510 |
2.5 L | Safebreak bottle |
Each for $217.87
|
|
||||
Description
- 100% anhydrous for analysis
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
2.5 (50g/L, H2O, 20°C) | |
64-19-7 | |
Colorless | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
1.05g/cm3 | |
4 to 19.9% | |
485°C | |
Glass bottle | |
<5ppm | |
Acetic Acid |
17°C | |
16°C to 118°C (1013hPa) | |
C2H4O2 | |
UN 2789 | |
602.9g/L | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
ACS/ISO/Reagent Ph. Eur. | |
2.5 L | |
39°C | |
Pungent/Vinegar-like | |
Liquid | |
15.4hPa(20°C) |
Safety and Handling
H226: Flammable liquid and vapor, H290: May be corrosive to metals, H314: Causes severe skin burns and eye damage,
P210: Keep away from heat, P280: Wear protective gloves/ protective clothing/ eye protection/ face protection, P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Corrosive
EINECSNumber : 200-580-7
RTECSNumber : AF1225000
Recommended Storage : Store at 15° to 25°C