missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, 6.0 N Solution, Spectrum™ Chemical
$76.76 - $76.76
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18608361
|
Spectrum Chemical Mfg Cor
A385500ML |
500 mL | Poly Bottle |
Each for $76.76
|
|
||||
Chemical Identifiers
64-19-7 |
Specifications
64-19-7,7732-18-5 | |
Chemical Solution | |
Poly Bottle |
6.0 N ± 0.06 N | |
500 mL |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.