missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid-Ammonium Acetate Buffer TS, (U.S.P. Test Solution), Spectrum™ Chemical
A-284, 64-19-7
$580.27 - $1357.10
Specifications
| CAS | 64-19-7,7732-18-5 |
|---|---|
| CAS Max % | 0.1%,99.9% |
| Grade | USP |
| Packaging | Amber Glass Bottle |
| Physical Form | Chemical Solution |