missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic acid-d, for NMR, 99.5 atom % D, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 464150250

Catalog No. AC464150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

40°C
1.07
1.3715
758-12-3
CH3CO2D
CC([O-])=O
59.05
NMR
25 mL
1.0700 g/mL
Acetic acid-d
C2H3O2
QTBSBXVTEAMEQO-UHFFFAOYSA-M
acetate
61.06
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye damage.

Causes severe skin burns and eye damage.

Flammable liquid and vapor.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

GHS Signal Word: Danger

EINECSNumber : 212-059-1

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only