Learn More
Acetic acid, Environmental Grade Plus, 99.4% min, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 38739AP
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetic acid | |
16.6°C | |
118.1°C | |
MFCD00036152 | |
14,55 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Environmental Plus | |
500 mL | |
Pungent | |
1.3721 |
2.5 | |
64-19-7 | |
C2H4O2 | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≤99.4% | |
1.049 g/mL | |
40°C (104°F) | |
Liquid |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
EINECSNumber : 200-580-7
TSCA : Yes
Recommended Storage : Ambient temperatures