missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic acid, glacial, 99.7+% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 039745AP

 View more versions of this product

Catalog No. AA39745AP


Explore available promotions

Description

Description

Good polar protic solvent, in its ester form it act as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetic acid
Glacial
64-19-7
(metals basis)
MFCD00036152
14,55
Miscible with water,ethanol,acetone,toluene and immiscible in carbon disulphide.
CC(O)=O
60.05
CHEBI:15366
≥99.9%
500 mL
Pungent
1.3721
2.5
16.6°C
118.1°C
C2H4O2
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
1.049 g/mL
40°C (104°F)
Liquid

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL

EINECSNumber : 200-580-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only