missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Glacial, 99%, Spectrum™ Chemical
A1720, 64-19-7, C2H4O2
Supplier: Spectrum Chemical Mfg Cor A172020LT
Description
Spectrum™ Chemical Acetic Acid, Glacial is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. A useful chemical reagent for the production of chemical compounds, Acetic acid has many uses. Its main use resides in the production of vinyl acetate monomer, but it is also used in the production of ester and acetic anhydride. In this case, in it′s Glacial form (undiluted), it is used as a polar protic solvent. As a solvent it is used in the purification by recrystallization of organic compounds and in the production of terephtalic acid.Specifications
64-19-7 | |
C2H4O2 | |
UN2789 | |
CC(O)=O | |
60.05 | |
Reagent | |
Poly Drum |
100% | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
99% | |
20 L |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
CC(O)=O |
C2H4O2 | |
MFCD00036152 | |
acetic acid |