Learn More
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036289K3
Description
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetic acid | |
Glacial | |
64-19-7 | |
99.7+% | |
MFCD00036152 | |
14,55 | |
Miscible with water. | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% | |
1.049 g/mL | |
40°C (104°F) | |
Liquid |
2.5 | |
16.6°C | |
118.1°C | |
C2H4O2 | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
ACS | |
2 L | |
Pungent | |
1.3721 |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
EINECSNumber : 200-580-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only