missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid Glacial ACS, MilliporeSigma™

Supplier:  Technidata AX00736

 View more versions of this product

Catalog No. MAX00736


Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
17°C
116°C to 118°C (1013hPa)
C2H4O2
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
6.5
1.05g/cm3 (20°C)
4 to 19.9%(V)
485°C
15.4 hPa (20°C)
Suprapur™
64-19-7
100%
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
ACS
500 mL
39.9°C (103.8°F)
Glass Bottle

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to 25°C

SDS
Documents

Documents

Promotions

Promotions