missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid Glacial ACS, MilliporeSigma™
Supplier: Technidata AX0073P5
Specifications
Acetic Acid | |
64-19-7 | |
C2H4O2 | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
6.5 | |
1.05g/cm3 (20°C) | |
4 to 19.9%(V) | |
Poly Coated Glass Bottle |
Suprapur™ | |
100% | |
CH3COOH | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
ACS | |
500 mL | |
485°C | |
15.4 hPa (20°C) |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
Recommended Storage : Store at +15°C to 25°C