Learn More
Acetic Acid, Glacial, ACS Grade, ≥ 99.7%, LabChem™
Supplier: LabChem LC1010036S
Specifications
Acetic Acid | |
17°C | |
118°C | |
Colorless | |
CH3COOH | |
UN2789 | |
Soluble in water | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% | |
1049kg/m3 | |
40°C | |
PVC Coated | |
≤0.0010% |
Glacial | |
64-19-7 | |
100 | |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
ACS | |
6 x 2.5 L | |
Vinegar | |
Liquid | |
1g/mL |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/spark/open flames/hot surfaces.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state, and federal regulations.
Danger
EINECSNumber : 607-002-00-6
Recommended Storage : Room Temperature