missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™
For DNA sequencing and other biological applications such as DNA/RNA electrophoresis
Supplier: Fisher BioReagents BP1185500
Description
For DNA sequencing and other biological applications such as DNA/RNA electrophoresis
Aldehyde-free Acetic Acid is suitable for DNA sequencing and other biological applications.Specifications
Acetic Acid, Glacial | |
Aldehyde-free | |
64-19-7 | |
Colorless | |
C2H4O2 | |
MFCD00036152 | |
15, 54 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Sequencing | |
500 mL | |
Liquid | |
1.52 kPa at 20°C |
2.5 | |
16°C | |
117°C | |
≥99.7 % | |
CH3COOH | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% | |
1.048g/cm³ | |
Glass | |
8ppm max. |