missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Glacial (HPLC), Fisher Chemical™
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
$316.58 - $316.58
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A35-500
|
Thermo Fisher Scientific
FLA35500 |
500 mL | Glass Bottle |
N/A
|
|
||||
Description
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1.0 AU max. at 255nm | |
16°C | |
117°C | |
Color (APHA) 10 | |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% (w/w) | |
1.048g/cm³ | |
Glass Bottle | |
0.001% | |
Acetic Acid, Glacial, 17.4N |
2.5 | |
64-19-7 | |
Colorless | |
≥99.7 w/w % CH3COOH | |
CH3COOH | |
2789 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
HPLC | |
500 mL | |
Liquid | |
1.52 kPa at 20°C |
Safety and Handling
DOTInformation : Class 8: Corrosive