missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Glacial (Optima™), Fisher Chemical™
Supplier: Seastar Chemicals Inc S020601TFIF01
Description
- Certificate of Analysis included with each bottle.
- Ideal for analysis by ICP-MS and high-resolution ICP-MS
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA.
Specifications
Acetic Acid | |
16°C | |
117°C | |
C2H4O2 | |
MFCD00036152 | |
15, 54 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
1.048g/cm³ | |
FEP Bottle | |
1.52 kPa at 20°C |
2.5 | |
64-19-7 | |
Colorless | |
CH3COOH | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
OPTIMA™ | |
250 mL | |
Liquid |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive